2,4-Dichloropyrimidine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5465-250G OR5465-100G OR5465-25G
APOSOR5465-250GEA 103 EUR
APOSOR5465-250G APOSOR5465-100G APOSOR5465-25G
2,4-Dichloropyrimidine 97%
2,4-Dichloropyrimidine
Formule: C₄H₂Cl₂N₂
Kookpunt: 101 °C/23mmHg
Smeltpunt: 57…61 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006061
CAS nummer: 3934-20-1
EINECS: 223-508-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR