4-Iodophenylboronic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid

OR5466-5G OR5466-1G
APOSOR5466-5GEA 42.4 EUR
APOSOR5466-5G APOSOR5466-1G
4-Iodophenylboronic acid 97%
4-Iodophenylboronic acid
Formule: C₆H₆BIO₂
MW: 247.83 g/mol
Smeltpunt: 322…326 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01319014
CAS nummer: 5122-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR