Furosemide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

OR5475-100G OR5475-25G OR5475-5G
APOSOR5475-100GEA 203 EUR
APOSOR5475-100G APOSOR5475-25G APOSOR5475-5G
Furosemide
Furosemide
Formule: C₁₂H₁₁ClN₂O₅S
MW: 330.74 g/mol
Smeltpunt: 220 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010549
CAS nummer: 54-31-9
EINECS: 200-203-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR