Pentan-3-ylhydrazine dihydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR55003-1G OR55003-5G
APOSOR55003-1GEA 177 EUR
APOSOR55003-1G APOSOR55003-5G
Pentan-3-ylhydrazine dihydrochloride
Pentan-3-ylhydrazine dihydrochloride
Formule: C₅H₁₄N₂
Smeltpunt: 60…80 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12401615
CAS nummer: 1211764-55-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR