1-Boc-5-cyanoindole-2-boronic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5550-1G OR5550-100MG
APOSOR5550-1GEA 261 EUR
APOSOR5550-1G APOSOR5550-100MG
1-Boc-5-cyanoindole-2-boronic acid 98%
1-Boc-5-cyanoindole-2-boronic acid
Formule: C₁₄H₁₅BN₂O₄
Smeltpunt: 260…270 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD04114578
CAS nummer: 475102-15-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR