(S)-4-Methyl-2-(hydroxymethyl)morpholine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR55501-1G OR55501-250MG
APOSOR55501-1GEA 303 EUR
APOSOR55501-1G APOSOR55501-250MG
(S)-4-Methyl-2-(hydroxymethyl)morpholine 95%
(S)-4-Methyl-2-(hydroxymethyl)morpholine
Formule: C₆H₁₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16294695
CAS nummer: 1159598-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR