4-Bromo-1-benzofuran-2-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR55703-1G OR55703-250MG OR55703-5G
APOSOR55703-1GEA 170 EUR
APOSOR55703-1G APOSOR55703-250MG APOSOR55703-5G
4-Bromo-1-benzofuran-2-carboxylic acid
4-Bromo-1-benzofuran-2-carboxylic acid
Formule: C₉H₅BrO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD22034498
CAS nummer: 1369143-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR