Ethyl-2-amino-5-oxazolecarboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5611-5G OR5611-1G OR5611-250MG
APOSOR5611-5GEA 80.9 EUR
APOSOR5611-5G APOSOR5611-1G APOSOR5611-250MG
Ethyl-2-amino-5-oxazolecarboxylate
Ethyl-2-amino-5-oxazolecarboxylate
Formule: C₆H₈N₂O₃
Smeltpunt: 151…153 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06657433
CAS nummer: 113853-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR