Indole-2-carbaldehyd

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5662-5G OR5662-1G OR5662-250MG
APOSOR5662-5GEA 103 EUR
APOSOR5662-5G APOSOR5662-1G APOSOR5662-250MG
Indole-2-carbaldehyd
Indole-2-carbaldehyd
Formule: C₉H₇NO
Smeltpunt: 138…142 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03001425
CAS nummer: 19005-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR