4-Bromo-N-(3-chloropropyl)benzenesulphonamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR5703-1G OR5703-5G
APOSOR5703-1GEA 121 EUR
APOSOR5703-1G APOSOR5703-5G
4-Bromo-N-(3-chloropropyl)benzenesulphonamide 98%
4-Bromo-N-(3-chloropropyl)benzenesulphonamide
Formule: C₉H₁₁BrClNO₂S
Smeltpunt: 88…90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01215119
CAS nummer: 98768-71-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR