3-Bromo-N-butylbenzenesulphonamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR5728-1G OR5728-5G
APOSOR5728-1GEA 125 EUR
APOSOR5728-1G APOSOR5728-5G
3-Bromo-N-butylbenzenesulphonamide 98%
3-Bromo-N-butylbenzenesulphonamide
Formule: C₁₀H₁₄BrNO₂S
Smeltpunt: 46…50 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07363822
CAS nummer: 871269-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR