Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5784-100G OR5784-25G OR5784-5G
APOSOR5784-100GEA 316 EUR
APOSOR5784-100G APOSOR5784-25G APOSOR5784-5G
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Formule: C13H13N3O4
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02947731
CAS nummer: 89392-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR