(2,4-Dichlorophenyl)acetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5793-100G OR5793-25G OR5793-500G
APOSOR5793-100GEA 36.6 EUR
APOSOR5793-100G APOSOR5793-25G APOSOR5793-500G
(2,4-Dichlorophenyl)acetic acid
(2,4-Dichlorophenyl)acetic acid
Formule: (C₆H₃Cl₂)CH₂COOH
Smeltpunt: 128…131 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004318
CAS nummer: 19719-28-9
EINECS: 243-248-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR