N-Benzyl-3-bromo-4-methylbenzenesulphonamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR5810-1G OR5810-5G
APOSOR5810-1GEA 172 EUR
APOSOR5810-1G APOSOR5810-5G
N-Benzyl-3-bromo-4-methylbenzenesulphonamide 98%
N-Benzyl-3-bromo-4-methylbenzenesulphonamide
Formule: C₁₄H₁₄BrNO₂S
Smeltpunt: 98…101 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07363832
CAS nummer: 850429-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR