L-Phenylalaninol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR5852-100G
APOSOR5852-100GEA 36.6 EUR
APOSOR5852-100G
L-Phenylalaninol 97%
L-Phenylalaninol
Formule: C₉H₁₃NO
Smeltpunt: 92…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004732
CAS nummer: 3182-95-4
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR