Isonicotinamidine hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR5939-1G OR5939-5G OR5939-25G
APOSOR5939-1GEA 22 EUR
APOSOR5939-1G APOSOR5939-5G APOSOR5939-25G
Isonicotinamidine hydrochloride
Isonicotinamidine hydrochloride
Formule: C₆H₇N₃·ClH
Smeltpunt: 263…265 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040385
CAS nummer: 42518-06-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR