2-Thiophenemethanol 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR59889-100G OR59889-25G OR59889-500G
APOSOR59889-100GEA 97.9 EUR
APOSOR59889-100G APOSOR59889-25G APOSOR59889-500G
2-Thiophenemethanol 99%
2-Thiophenemethanol
Formule: C₅H₆OS
Kookpunt: 206…207 °C
Dichtheid: 1,205 g/cm³ (25 °C)
Vlampunt: 110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005454
CAS nummer: 636-72-6
EINECS: 211-264-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR