Cyclopropylacetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR59893-25G OR59893-5G
APOSOR59893-25GEA 234 EUR
APOSOR59893-25G APOSOR59893-5G
Cyclopropylacetic acid
Cyclopropylacetic acid
Formule: C₅H₈O₂
Kookpunt: 189…191 °C
Dichtheid: 1,076 g/cm³ (25 °C)
Vlampunt: 93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041544
CAS nummer: 5239-82-7
UN: 3265
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR