5-Bromo-4-methyl-2-phenyl-1,3-oxazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR59975-5G OR59975-500MG OR59975-2.5G
APOSOR59975-5GEA 951 EUR
APOSOR59975-5G APOSOR59975-500MG APOSOR59975-2.5G
5-Bromo-4-methyl-2-phenyl-1,3-oxazole
5-Bromo-4-methyl-2-phenyl-1,3-oxazole
Formule: C₁₀H₈BrNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11109319
CAS nummer: 21354-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR