4-Iodobenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR60044-5G OR60044-10G OR60044-25G
APOSOR60044-5GEA 26.3 EUR
APOSOR60044-5G APOSOR60044-10G APOSOR60044-25G
4-Iodobenzyl alcohol
4-Iodobenzyl alcohol
Formule: C₇H₇IO
Smeltpunt: 72…75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01732720
CAS nummer: 18282-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR