6-Isoquinolineboronic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR60071-1G OR60071-250MG
APOSOR60071-1GEA 238 EUR
APOSOR60071-1G APOSOR60071-250MG
6-Isoquinolineboronic acid 95%
6-Isoquinolineboronic acid
Formule: C₉H₈BNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10697418
CAS nummer: 899438-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR