(S)-4-Hydroxypyrrolidine-2-one

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (4S)-(-)-beta-Hydroxy-gamma-butyrolactam, (S)-4-Hydroxy-2-pyrrolidone, (S)-(-)-4-Hydroxy-2-pyrrolidinone

OR60184-25G
APOSOR60184-25GEA 144 EUR
APOSOR60184-25G
(S)-4-Hydroxypyrrolidine-2-one
(S)-4-Hydroxypyrrolidine-2-one
Formule: C₄H₇NO₂
Smeltpunt: 156…159 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00273370
CAS nummer: 68108-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR