Pyrrole-2-carbonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR60193-25G OR60193-100G
APOSOR60193-25GEA 73 EUR
APOSOR60193-25G APOSOR60193-100G
Pyrrole-2-carbonitrile 98%
Pyrrole-2-carbonitrile
Formule: C₅H₄N₂
Kookpunt: 92…94 °C/2mmHg
Dichtheid: 1,086 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00234061
CAS nummer: 4513-94-4
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR