5-Bromo-2-[(methylamino)methyl]pyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR6090-1G OR6090-250MG
APOSOR6090-1GEA 463 EUR
APOSOR6090-1G APOSOR6090-250MG
5-Bromo-2-[(methylamino)methyl]pyridine 95%
5-Bromo-2-[(methylamino)methyl]pyridine
Formule: C₇H₉BrN₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09065029
CAS nummer: 915707-70-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR