2-Amino-3-chloropyrazine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR6127-5G OR6127-1G OR6127-25G
APOSOR6127-5GEA 22 EUR
APOSOR6127-5G APOSOR6127-1G APOSOR6127-25G
2-Amino-3-chloropyrazine 97%
2-Amino-3-chloropyrazine
Formule: C₄H₄ClN₃
Kookpunt: 248 °C
Smeltpunt: 167…171 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04114305
CAS nummer: 6863-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR