Pinacol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR61505-100G OR61505-25G OR61505-500G
APOSOR61505-100GEA 35.1 EUR
APOSOR61505-100G APOSOR61505-25G APOSOR61505-500G
Pinacol
Pinacol
Formule: C₆H₁₄O₂
Kookpunt: 174 °C
Smeltpunt: 40…43 °C
Vlampunt: 77 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004462
CAS nummer: 76-09-5
EINECS: 200-933-5
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR