2,6-Diaminopurine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine

OR6157-1G OR6157-5G OR6157-10G OR6157-25G
APOSOR6157-1GEA 14.8 EUR
APOSOR6157-1G APOSOR6157-5G APOSOR6157-10G APOSOR6157-25G
2,6-Diaminopurine 98%
2,6-Diaminopurine
Formule: C₅H₆N₆
MW: 150.14 g/mol
Smeltpunt: >300 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00213668
CAS nummer: 1904-98-9
EINECS: 217-605-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR