4-(1H-Pyrazol-4-yl)benzoic acid 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR6230-250MG
APOSOR6230-250MGEA 87.6 EUR
APOSOR6230-250MG
4-(1H-Pyrazol-4-yl)benzoic acid 95+%
4-(1H-Pyrazol-4-yl)benzoic acid
Formule: C₁₀H₈N₂O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06803014
CAS nummer: 1017794-47-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR