1,2-Dichloro-4-iodobenzene 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3,4-Dichloroiodobenzene

OR6486-100G OR6486-250G OR6486-25G OR6486-500G
APOSOR6486-100GEA 49.7 EUR
APOSOR6486-100G APOSOR6486-250G APOSOR6486-25G APOSOR6486-500G
1,2-Dichloro-4-iodobenzene 99%
1,2-Dichloro-4-iodobenzene
Formule: C₆H₃Cl₂I
MW: 272.9 g/mol
Kookpunt: 116…118 °C/12mmHg
Smeltpunt: 27…29 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00019014
CAS nummer: 20555-91-3
EINECS: 243-874-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR