3-Amino-2-chloropyridine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

OR6543-100G OR6543-25G OR6543-5G OR6543-1G OR6543-500G
APOSOR6543-100GEA 0 EUR
APOSOR6543-100G APOSOR6543-25G APOSOR6543-5G APOSOR6543-1G APOSOR6543-500G
3-Amino-2-chloropyridine 98%
3-Amino-2-chloropyridine
Formule: C₅H₅ClN₂
MW: 128.56 g/mol
Kookpunt: 130…134 °C/22mmHg
Smeltpunt: 76…78 °C
Vlampunt: 185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006238
CAS nummer: 6298-19-7
EINECS: 228-572-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR