N-(4,6-Dimethylpyrimidin-2-yl)-2-aminobenzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR6554-250MG
APOSOR6554-250MGEA 74.6 EUR
APOSOR6554-250MG
N-(4,6-Dimethylpyrimidin-2-yl)-2-aminobenzoic acid
N-(4,6-Dimethylpyrimidin-2-yl)-2-aminobenzoic acid
Formule: C₁₃H₁₃N₃O₂
Smeltpunt: 252…254 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00203260
CAS nummer: 17174-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR