1-Bromo-4-chloro-3-nitrobenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Bromo-1-chloro-2-nitrobenzene, 5-Bromo-2-chloronitrobenzene

OR6626-100G OR6626-25G OR6626-5G
APOSOR6626-100GEA 71.6 EUR
APOSOR6626-100G APOSOR6626-25G APOSOR6626-5G
1-Bromo-4-chloro-3-nitrobenzene
1-Bromo-4-chloro-3-nitrobenzene
Formule: C₆H₃BrClNO₂
MW: 236.4500 g/mol
Smeltpunt: 70…72 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05863216
CAS nummer: 16588-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR