4-(Piperidin-1-ylmethyl)benzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR6740-1G OR6740-250MG OR6740-5G
APOSOR6740-1GEA 220 EUR
APOSOR6740-1G APOSOR6740-250MG APOSOR6740-5G
4-(Piperidin-1-ylmethyl)benzaldehyde 95%
4-(Piperidin-1-ylmethyl)benzaldehyde
Formule: C₁₃H₁₇NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07772862
CAS nummer: 471929-86-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR