4-(2-Methylthiazol-4-yl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR6745-1G OR6745-10G OR6745-5G
APOSOR6745-1GEA 150 EUR
APOSOR6745-1G APOSOR6745-10G APOSOR6745-5G
4-(2-Methylthiazol-4-yl)benzoic acid
4-(2-Methylthiazol-4-yl)benzoic acid
Formule: C₁₁H₉NO₂S
Smeltpunt: 263…268 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02225088
CAS nummer: 294620-60-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR