Diethyl (cyanomethyl)phosphonate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR6821-100G OR6821-25G OR6821-250G
APOSOR6821-100GEA 58.4 EUR
APOSOR6821-100G APOSOR6821-25G APOSOR6821-250G
Diethyl (cyanomethyl)phosphonate 98%
Diethyl (cyanomethyl)phosphonate
Formule: C₆H₁₂NO₃P
Kookpunt: 101…102 °C/0,4mmHg
Dichtheid: 1,095 g/cm³ (25 °C)
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001893
CAS nummer: 2537-48-6
EINECS: 219-806-0
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR