1-(2-Pyridyl)piperazine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR6871-100G OR6871-1KG OR6871-250G
APOSOR6871-100GEA 141 EUR
APOSOR6871-100G APOSOR6871-1KG APOSOR6871-250G
1-(2-Pyridyl)piperazine 98%
1-(2-Pyridyl)piperazine
Formule: C₉H₁₃N₃
Kookpunt: 120…122 °C/2mmHg
Dichtheid: 1,072 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006216
CAS nummer: 34803-66-2
EINECS: 252-220-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR