4-Chloro-6-nitro-o-toluidine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Chloro-2-methyl-6-nitroaniline

OR6914-250MG OR6914-5G
APOSOR6914-250MGEA 22 EUR
APOSOR6914-250MG APOSOR6914-5G
4-Chloro-6-nitro-o-toluidine 98%
4-Chloro-6-nitro-o-toluidine
Formule: C₇H₇ClN₂O₂
MW: 186.60 g/mol
Smeltpunt: 121…123 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01320687
CAS nummer: 62790-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR