m-Toluic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-Methylbenzoic acid

OR70175-500G OR70175-100G
APOSOR70175-500GEA 24.9 EUR
APOSOR70175-500G APOSOR70175-100G
m-Toluic acid
m-Toluic acid
Formule: C₈H₈O₂
MW: 136.15 g/mol
Kookpunt: 263
Vlampunt: 150
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002523
CAS nummer: 99-04-7
Merck Index: 12,09673

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR