2-Amino-5-chloro-3-iodobenzoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR70216-1G OR70216-5G OR70216-250MG
APOSOR70216-1GEA 24.9 EUR
APOSOR70216-1G APOSOR70216-5G APOSOR70216-250MG
2-Amino-5-chloro-3-iodobenzoic acid 95%
2-Amino-5-chloro-3-iodobenzoic acid
Formule: C₇H₅ClINO₂
Smeltpunt: 246…247 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07780627
CAS nummer: 64724-23-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR