8-Bromo-4-cyanoquinoline-n-oxide ≥95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR70311-250MG OR70311-1G
APOSOR70311-250MGEA 228 EUR
APOSOR70311-250MG APOSOR70311-1G
8-Bromo-4-cyanoquinoline-n-oxide ≥95%
8-Bromo-4-cyanoquinoline-n-oxide
Formule: C₁₀H₅BrN₂O
MDL Nummer: MFCD32862645
CAS nummer: 2640734-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR