3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7084-1G OR7084-250MG
APOSOR7084-1GEA 176 EUR
APOSOR7084-1G APOSOR7084-250MG
3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride
3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride
Formule: C₈H₁₅N₃·2ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07366477
CAS nummer: 423176-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR