2-N-methylamino-6-bromobenzoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR71110-250MG OR71110-1G
APOSOR71110-250MGEA 125 EUR
APOSOR71110-250MG APOSOR71110-1G
2-N-methylamino-6-bromobenzoic acid 95%
2-N-methylamino-6-bromobenzoic acid
Formule: C₈H₈BrNO₂
MDL Nummer: MFCD28356796
CAS nummer: 1369866-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR