N,N-Dimethyl 2-boronobenzenesulphonamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7212-1G OR7212-5G
APOSOR7212-1GEA 129 EUR
APOSOR7212-1G APOSOR7212-5G
N,N-Dimethyl 2-boronobenzenesulphonamide
N,N-Dimethyl 2-boronobenzenesulphonamide
Formule: C₈H₁₂BNO₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD04039023
CAS nummer: 178432-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR