2-Methoxy-5-pyridineboronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7235-25G OR7235-5G OR7235-100G
APOSOR7235-25GEA 55.5 EUR
APOSOR7235-25G APOSOR7235-5G APOSOR7235-100G
2-Methoxy-5-pyridineboronic acid
2-Methoxy-5-pyridineboronic acid
Formule: C₆H₈BNO₃
Smeltpunt: 135…140 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02093044
CAS nummer: 163105-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR