Apigenin 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR7265T-25G OR7265T-5G OR7265T-100G
APOSOR7265T-25GEA 117 EUR
APOSOR7265T-25G APOSOR7265T-5G APOSOR7265T-100G
Apigenin 97%
Apigenin

A MAP kinase inhibitor. Also inhibits the proliferation of malignant tumor cells.

Formule: C₁₅H₁₀O₅
Smeltpunt: 300 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006831
CAS nummer: 520-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR