3-Bromo-5-chloroaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR7300-10G OR7300-5G OR7300-25G
APOSOR7300-10GEA 45.4 EUR
APOSOR7300-10G APOSOR7300-5G APOSOR7300-25G
3-Bromo-5-chloroaniline 98%
3-Bromo-5-chloroaniline
Formule: C₆H₅BrClN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06797804
CAS nummer: 96558-78-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR