3-(2-Furyl)benzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7403-1G OR7403-250MG OR7403-50MG
APOSOR7403-1GEA 236 EUR
APOSOR7403-1G APOSOR7403-250MG APOSOR7403-50MG
3-(2-Furyl)benzonitrile
3-(2-Furyl)benzonitrile
Formule: C₁₁H₇NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04039075
CAS nummer: 112598-77-3
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR