Silver bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7534-5G OR7534-100G OR7534-25G
APOSOR7534-5GEA 22 EUR
APOSOR7534-5G APOSOR7534-100G APOSOR7534-25G
Silver bromide
Silver bromide
Formule: AgBr
Smeltpunt: 432 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003398
CAS nummer: 7785-23-1
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR