5-Bromo-3-methylindole 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7820-1G OR7820-25G OR7820-5G
APOSOR7820-1GEA 121 EUR
APOSOR7820-1G APOSOR7820-25G APOSOR7820-5G
5-Bromo-3-methylindole 98%
5-Bromo-3-methylindole
Formule: C₉H₈BrN
Smeltpunt: 76…82 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD04038746
CAS nummer: 10075-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR