4-(tert-Butoxycarbonyl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7829-5G
APOSOR7829-5GEA 97.9 EUR
APOSOR7829-5G
4-(tert-Butoxycarbonyl)benzoic acid
4-(tert-Butoxycarbonyl)benzoic acid
Formule: C₁₂H₁₄O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04037901
CAS nummer: 20576-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR